Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amikacin, MP Biomedicals™

Catalog No. MP21503422 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21503422 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21503422 Supplier MP Biomedicals, Inc Supplier No. 02150342.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to treat different types of bacterial infections. Amikacin, MP Biomedicals is an aminoglycoside antibiotic most often used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Ente

Amikacin works by binding to the bacterial 30S ribosomal subunit, inducing misreading of mRNA and leaving the bacterium unable to synthesize proteins vital to its growth. It interferes with the translocation of tRNA from the A-site to the P-site. Amikacin must be given by the intravenous or intramuscular route; it is not absorbed orally.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37517-28-5
Molecular Formula C22H43N5O13
Molecular Weight (g/mol) 585.608
InChI Key LKCWBDHBTVXHDL-RMDFUYIESA-N
Synonym 1-N-[L(-)-4-Amino-2-hydroxy-butyryl] kanamycin A, O-3-Amino-3-deoxy-a -D-glucopyranosyl-(1 ---> 6)-O-6-amino-6-deoxy-a-D-glucopyranosyl-(1 ---> 4)]- N1 -(4-amino-2-hydroxy-1-oxobutyl)-2-deoxy-D-streptamine, Lukadin
PubChem CID 37768
ChEBI CHEBI:2637
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide
SMILES C1C(C(C(C(C1NC(=O)C(CCN)O)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(C(C(C(O3)CN)O)O)O)N

Specifications

Physical Form Powder
pH 9.5 to 11.5 (1% aq. soln.)
Potency ca. 900μg/mg
Color White to Light Gray
Quantity 250 mg
Optical Rotation 100.5°
Boiling Point 981.8°C (760mmHg)
Solubility Information Sparingly soluble in water.
Formula Weight 585.6
Grade Not Available
Recommended Storage Store at 4°C, desssicated.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.