Learn More
Medchemexpress LLC Amikacin sulfate | 149022-22-0 | MFCD00167477 | 99.1% | 762.14 | C22H45N5O17S | 100 MG
SDP
Catalog No. 5000242303
Encompass_Preferred
Amikacin sulfate is an aminoglycoside antibiotic supplied as a sulfate salt for laboratory and research use. It is bactericidal and inhibits bacterial protein synthesis by binding to the ribosomal subunits, showing activity against many Gram-negative organisms and certain mycobacteria and Nocardia species. Often used in microbiology and pharmacology studies, it is supplied with high purity for analytical and experimental applications.
  • Bactericidal aminoglycoside that inhibits protein synthesis.
  • Acts on 30S and 50S bacterial ribosomal subunits.
  • Active against many Gram-negative bacteria, including resistant strains.
  • Supplied as a sulfate salt suitable for research use.
  • High purity (99.1%) for analytical and experimental applications.
Catalog No. 50-002-42303 Supplier Medchemexpress LLC Supplier No. HY107813100MG
Amikacin sulfate is an aminoglycoside antibiotic supplied as a sulfate salt for laboratory and research use. It is bactericidal and inhibits bacterial protein synthesis by binding to the ribosomal subunits, showing activity against many Gram-negative organisms and certain mycobacteria and Nocardia species. Often used in microbiology and pharmacology studies, it is supplied with high purity for analytical and experimental applications.
  • Bactericidal aminoglycoside that inhibits protein synthesis.
  • Acts on 30S and 50S bacterial ribosomal subunits.
  • Active against many Gram-negative bacteria, including resistant strains.
  • Supplied as a sulfate salt suitable for research use.
  • High purity (99.1%) for analytical and experimental applications.