Learn More
Medchemexpress LLC Amikacin sulfate 50mg | 149022-22-0 | 50MG
SDP
Catalog No. 5000242304
Encompass_Preferred
Amikacin sulfate (BAY 41-6551 sulfate) is an aminoglycoside antibiotic and a semisynthetic analog of kanamycin Amikacin sulfate is bactericidal acting directly on the 30S and 50S bacerial ribosomal subunits to inhibit protein synthesis Amikacin sulfate is very active against most Gram-negative bacteria including gentamicin- and tobramycin-resistant strains Amikacin sulfate also inhibits the infections caused by susceptible Nocardia and nontuberculous mycobacteria[1 [2
Catalog No. 50-002-42304 Supplier Medchemexpress LLC Supplier No. HY10781350MG
Amikacin sulfate (BAY 41-6551 sulfate) is an aminoglycoside antibiotic and a semisynthetic analog of kanamycin Amikacin sulfate is bactericidal acting directly on the 30S and 50S bacerial ribosomal subunits to inhibit protein synthesis Amikacin sulfate is very active against most Gram-negative bacteria including gentamicin- and tobramycin-resistant strains Amikacin sulfate also inhibits the infections caused by susceptible Nocardia and nontuberculous mycobacteria[1 [2