Learn More
Selleck Chemical LLC Amikacin Sulfate Salt S5330-100mg
SDP
Catalog No. 503109478
Encompass_Preferred
Amikacin (BAY 41-6551) sulfate a semisynthetic analog of kanamycin is an aminoglycoside antibiotic which is very active against most gram-negative bacteria including gentamicin- and tobramycin-resistant strains
Catalog No. 50-310-9478 Supplier Selleck Chemical LLC Supplier No. S5330100MG
Amikacin (BAY 41-6551) sulfate a semisynthetic analog of kanamycin is an aminoglycoside antibiotic which is very active against most gram-negative bacteria including gentamicin- and tobramycin-resistant strains