Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amikacin Sulfate, USP Grade, G-Biosciences
SCI_ED

Catalog No. S139839
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139839 25 mg
S139725 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139839 Supplier G-Biosciences Supplier No. RC881
Add to Cart
Add to Cart

Amikacin sulfate (disulfate) is a broad-spectrum aminoglycoside antibiotic derived from kanamycin A.

It is effective against gram-negative bacteria such as Pseudomonas aeruginosa and a variety of enteric species. It is also effective against Mycobacterium tuberculosis.

Specifications

Boiling Point 632.23°C
Quantity 25 mg
Molecular Weight (g/mol) 781.76 g/mol
Percent Purity >95%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.