Learn More
Medchemexpress LLC Amilo-5MER TFA (5-MP TFA) | 99.6% | 576.66 (free base) | 5 MG
SDP
Catalog No. 5000424116
Encompass_Preferred
Amilo-5MER TFA (5-MP) is an orally active and selective Serum Amyloid A (SAA) inhibitor. It specifically inhibits the release of pro-inflammatory cytokines IL-6 and IL-1β from SAA-activated cells. This compound has shown the ability to reduce chronic inflammation and alleviate symptoms associated with diseases like rheumatoid arthritis (RA), inflammatory bowel disease (IBD), and multiple sclerosis (MS). It is considered promising for research in autoimmune and chronic inflammatory diseases.
  • Orally active and selective Serum Amyloid A (SAA) inhibitor.
  • Specifically inhibits the release of pro-inflammatory cytokines IL-6 and IL-1β from SAA-activated cells.
  • Reduces chronic inflammation.
  • Relieves symptoms of diseases such as rheumatoid arthritis (RA), inflammatory bowel disease (IBD), and multiple sclerosis (MS).
  • Promising for research of autoimmune diseases and chronic inflammatory diseases.
Catalog No. 50-004-24116 Supplier Medchemexpress LLC Supplier No. HYP10935A5MG
Amilo-5MER TFA (5-MP) is an orally active and selective Serum Amyloid A (SAA) inhibitor. It specifically inhibits the release of pro-inflammatory cytokines IL-6 and IL-1β from SAA-activated cells. This compound has shown the ability to reduce chronic inflammation and alleviate symptoms associated with diseases like rheumatoid arthritis (RA), inflammatory bowel disease (IBD), and multiple sclerosis (MS). It is considered promising for research in autoimmune and chronic inflammatory diseases.
  • Orally active and selective Serum Amyloid A (SAA) inhibitor.
  • Specifically inhibits the release of pro-inflammatory cytokines IL-6 and IL-1β from SAA-activated cells.
  • Reduces chronic inflammation.
  • Relieves symptoms of diseases such as rheumatoid arthritis (RA), inflammatory bowel disease (IBD), and multiple sclerosis (MS).
  • Promising for research of autoimmune diseases and chronic inflammatory diseases.