Learn More
Medchemexpress LLC Amilo-5MER TFA | 99.6% | 576.66 (free base) | 1 MG
SDP
Catalog No. 5000422776
Encompass_Preferred
Amilo-5MER (5-MP) TFA is an orally active and selective Serum Amyloid A (SAA) inhibitor. Amilo-5MER TFA specifically inhibits the release of pro-inflammatory cytokines IL-6 and IL-1β from SAA-activated cells. Amilo-5MER TFA reduces chronic inflammation and relieves symptoms of diseases such as rheumatoid arthritis (RA), inflammatory bowel disease (IBD), and multiple sclerosis (MS). Amilo-5MER TFA is promising for research of autoimmune diseases and chronic inflammatory diseases.
  • Orally active and selective serum amyloid A (SAA) inhibitor.
  • Specifically inhibits the release of pro-inflammatory cytokines IL-6 and IL-1β from SAA-activated cells.
  • Reduces chronic inflammation.
  • Relieves symptoms of diseases such as rheumatoid arthritis (RA), inflammatory bowel disease (IBD), and multiple sclerosis (MS).
  • Promising for research of autoimmune diseases and chronic inflammatory diseases.
Catalog No. 50-004-22776 Supplier Medchemexpress LLC Supplier No. HYP10935A1MG
Amilo-5MER (5-MP) TFA is an orally active and selective Serum Amyloid A (SAA) inhibitor. Amilo-5MER TFA specifically inhibits the release of pro-inflammatory cytokines IL-6 and IL-1β from SAA-activated cells. Amilo-5MER TFA reduces chronic inflammation and relieves symptoms of diseases such as rheumatoid arthritis (RA), inflammatory bowel disease (IBD), and multiple sclerosis (MS). Amilo-5MER TFA is promising for research of autoimmune diseases and chronic inflammatory diseases.
  • Orally active and selective serum amyloid A (SAA) inhibitor.
  • Specifically inhibits the release of pro-inflammatory cytokines IL-6 and IL-1β from SAA-activated cells.
  • Reduces chronic inflammation.
  • Relieves symptoms of diseases such as rheumatoid arthritis (RA), inflammatory bowel disease (IBD), and multiple sclerosis (MS).
  • Promising for research of autoimmune diseases and chronic inflammatory diseases.