Learn More
Enzo Life Sciences Amiloride, 5-(N-Ethyl-N-isopropyl)-, (5mg), CAS Number: 1154-25-2
SDP
Catalog No. NC1049834
Encompass_Preferred
Na+/H+ antiporter inhibitor. Selective inhibitor of the Na /H antiporter. Alternative Name: 5-(N-Ethyl-N-isopropyl)amiloride, EIPA. Formula: C11H18ClN7O. MW: 299.8. CAS Number: 1154-25-2. Purity: ≥98% (TLC). Appearance: Off-white to yellow solid. Solubility: Soluble in methanol, DMSO or dilute aqueous acid; insoluble in water. Long Term Storage: +4°C
Catalog No. NC1049834 Supplier Enzo Life Sciences Supplier No. ALX550266M005
Na+/H+ antiporter inhibitor. Selective inhibitor of the Na /H antiporter. Alternative Name: 5-(N-Ethyl-N-isopropyl)amiloride, EIPA. Formula: C11H18ClN7O. MW: 299.8. CAS Number: 1154-25-2. Purity: ≥98% (TLC). Appearance: Off-white to yellow solid. Solubility: Soluble in methanol, DMSO or dilute aqueous acid; insoluble in water. Long Term Storage: +4°C