Learn More
Selleck Chemical LLC Amiloride HCl S1811-1g
SDP
Supplier:  Selleck Chemical LLC S18111G
Encompass_Preferred
Amiloride HCl is a selective T-type calcium channel blocker an epithelial sodium channel blocker and a selective inhibitor of urokinase plasminogen activator (uPA)(Ki 7 M)
Catalog No. 50-310-9030