Learn More
Selleck Chemical LLC Amiloride HCl S1811-10mM/1mL
SDP
Catalog No. 503110792
Encompass_Preferred
Amiloride HCl is a selective T-type calcium channel blocker an epithelial sodium channel blocker and a selective inhibitor of urokinase plasminogen activator (uPA)(Ki 7 M)
Catalog No. 50-311-0792 Supplier Selleck Chemical LLC Supplier No. S181110MM/1ML
Add to Cart
Add to Cart
Amiloride HCl is a selective T-type calcium channel blocker an epithelial sodium channel blocker and a selective inhibitor of urokinase plasminogen activator (uPA)(Ki 7 M)