Learn More
Medchemexpress LLC Amiloride hydrochloride dihydrate | 17440-83-4 | 99.9% | 100 MG
SDP
Catalog No. 501970103
Encompass_Preferred
Amiloride hydrochloride dihydrate is an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA). It also acts as a blocker of polycystin-2 (PC2; TRPP2) channel. This product is for research use only and is not sold to patients.
  • Inhibitor of epithelial sodium channel (ENaC)
  • Inhibitor of urokinase-type plasminogen activator receptor (uTPA)
  • Blocker of polycystin-2 (PC2; TRPP2) channel
Catalog No. 50-197-0103 Supplier Medchemexpress LLC Supplier No. HYB0285B100MG
Amiloride hydrochloride dihydrate is an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA). It also acts as a blocker of polycystin-2 (PC2; TRPP2) channel. This product is for research use only and is not sold to patients.
  • Inhibitor of epithelial sodium channel (ENaC)
  • Inhibitor of urokinase-type plasminogen activator receptor (uTPA)
  • Blocker of polycystin-2 (PC2; TRPP2) channel