Learn More
Medchemexpress LLC Amiloride hydrochloride | 2016-88-8 | 99.8% | 25 MG
SDP
Catalog No. 5000450997
Encompass_Preferred
Amiloride hydrochloride is an analytical standard intended for research and analytical applications. It acts as an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA), and is also identified as a blocker of the polycystin-2 (PC2; TRPP2) channel. This compound is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Used as an analytical standard
  • Inhibits epithelial sodium channel (ENaC)
  • Inhibits urokinase-type plasminogen activator receptor (uTPA)
  • Blocks polycystin-2 (PC2; TRPP2) channel
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-50997 Supplier Medchemexpress LLC Supplier No. 25MGHYB0285AR
Amiloride hydrochloride is an analytical standard intended for research and analytical applications. It acts as an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA), and is also identified as a blocker of the polycystin-2 (PC2; TRPP2) channel. This compound is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Used as an analytical standard
  • Inhibits epithelial sodium channel (ENaC)
  • Inhibits urokinase-type plasminogen activator receptor (uTPA)
  • Blocks polycystin-2 (PC2; TRPP2) channel
  • Suitable for HPLC, GC, and MS experiments