Learn More
Medchemexpress LLC Amiloride hydrochloride | 2016-88-8 | MFCD03703482 | 99.8% | 5 MG
SDP
Catalog No. 5000405970
Encompass_Preferred
Amiloride hydrochloride is an analytical standard intended for research and analytical applications. It serves as an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA), and acts as a blocker of the polycystin-2 (PC2; TRPP2) channel. This product is suitable for various methodological research experiments.
  • Used as an analytical standard
  • Suitable for qualitative, quantitative, and methodological research
  • Applied in HPLC, GC, and MS experiments
  • Functions as an inhibitor of ENaC and uTPA
  • Acts as a blocker of polycystin-2 (PC2; TRPP2) channel
Catalog No. 50-004-05970 Supplier Medchemexpress LLC Supplier No. HYB0285AR5MG
Amiloride hydrochloride is an analytical standard intended for research and analytical applications. It serves as an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA), and acts as a blocker of the polycystin-2 (PC2; TRPP2) channel. This product is suitable for various methodological research experiments.
  • Used as an analytical standard
  • Suitable for qualitative, quantitative, and methodological research
  • Applied in HPLC, GC, and MS experiments
  • Functions as an inhibitor of ENaC and uTPA
  • Acts as a blocker of polycystin-2 (PC2; TRPP2) channel