Learn More
Medchemexpress LLC Amiloride hydrochloride dihydrate | 17440-83-4 | MFCD00211292 | 99.9% | 1 ML
SDP
Catalog No. 5000408718
Encompass_Preferred
Amiloride hydrochloride dihydrate is an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA). It also acts as a blocker of polycystin-2 (PC2; TRPP2) channel.
  • Inhibits epithelial sodium channel (ENaC)
  • Inhibits urokinase-type plasminogen activator receptor (uTPA)
  • Blocks polycystin-2 (PC2; TRPP2) channel
Catalog No. 50-004-08718 Supplier Medchemexpress LLC Supplier No. HYB0285B10MM/1ML
Amiloride hydrochloride dihydrate is an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA). It also acts as a blocker of polycystin-2 (PC2; TRPP2) channel.
  • Inhibits epithelial sodium channel (ENaC)
  • Inhibits urokinase-type plasminogen activator receptor (uTPA)
  • Blocks polycystin-2 (PC2; TRPP2) channel