Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amiloride hydrochloride, For HPLC analysis, 99.4%, MP Biomedicals™

Catalog No. ICN15353750 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15353750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15353750 Supplier MP Biomedicals, Inc Supplier No. 0215353750

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2016-88-8
Molecular Formula C6H9Cl2N7O
Molecular Weight (g/mol) 266.09
MDL Number MFCD00211292,MFCD03703482
InChI Key ACHKKGDWZVCSNH-UHFFFAOYSA-N
Synonym amiloride hydrochloride, amiloride hcl, amiloride chloride, amiloride hydrochloride anhydrous, ccris 5959, amilorida hydrochloride, amiloride, hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide monohydrochloride
PubChem CID 16230
ChEBI CHEBI:84743
IUPAC Name hydrogen 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide chloride
SMILES [H+].[Cl-].NC(N)=NC(=O)C1=NC(Cl)=C(N)N=C1N

Specifications

Color Yellow
Assay Percent Range 99.4%
Quantity 50 mg
Formula Weight 266.1
Physical Form Solid
Chemical Name or Material Amiloride hydrochloride, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.