Learn More
Cayman Chemical Amiloride (hydrochloride), 2016-88-8, 1 g
SDP
Supplier:  Cayman Chemical 144091
Encompass_Preferred
Molecular Formula C6H8ClN7O • HCl, Purity ≥98%, Formula Weight 266.1, Synonyms: 3,5-diamino-N-(aminoiminomethyl)-6-chloro-2-pyrazinecarboxamide, monohydrochloride, MK-870; A potassium-sparing diuretic that promotes the loss of sodium and water from the body by directly blocking epithelial sodium channels (ENaCs); also blocks Na+/H+ exchangers (NHE-1) in the heart and acid-sensing ion channels (ASICs), which are involved in nociceptor responses to pH.
Catalog No. NC1037879