Learn More
TARGETMOL CHEMICALS INC AMINAFTONE 5MG
SDP
Catalog No. 502581303
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Aminaftone a derivative of 4-aminobenzoic acid inhibits endothelin-1 (ET-1) production and can be used to study hypertension and systemic sclerosis. purity: 99%
Catalog No. 50-258-1303 Supplier TARGETMOL CHEMICALS INC Supplier No. T142195MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Aminaftone a derivative of 4-aminobenzoic acid inhibits endothelin-1 (ET-1) production and can be used to study hypertension and systemic sclerosis. purity: 99%