Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LUNA NANOTECH INC Amine Coated Magnetic Beads, 500 Nm, 10 ML

Catalog No. 502566982
Encompass_Preferred

500 nm diameter magnetic beads with Amine (NH2) functionalized surface.
10 mL @ 10 mg/mL

Features:
- Conjugation Ready: NH2 groups on the magnetic beads can be reacted with activated carboxyl groups to covalently load proteins, small molecules, or DNA / RNA onto the beads.
- High monodispersity.
- Stable in high salt conditions.
Fast response to a magnet.

Applications:
- Magnetic separation of DNA, RNA, proteins, antibodies, cells and small molecules from complex samples.
- Carboxyl containing proteins, small molecules, DNA / RNA can be covalently attached to the amine groups on the bead surface through EDC/NHS reactions.

Catalog No. 50-256-6982 Supplier LUNA NANOTECH INC Supplier No. MGBNH21010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

500 nm diameter magnetic beads with Amine (NH2) functionalized surface.
10 mL @ 10 mg/mL

Features:
- Conjugation Ready: NH2 groups on the magnetic beads can be reacted with activated carboxyl groups to covalently load proteins, small molecules, or DNA / RNA onto the beads.
- High monodispersity.
- Stable in high salt conditions.
Fast response to a magnet.

Applications:
- Magnetic separation of DNA, RNA, proteins, antibodies, cells and small molecules from complex samples.
- Carboxyl containing proteins, small molecules, DNA / RNA can be covalently attached to the amine groups on the bead surface through EDC/NHS reactions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.