Learn More
LUNA NANOTECH INC Amine-Functionlzd Dextran 40Kda
Catalog No. NC3593155
Encompass_Preferred
Dextran Size: 40 kDa
Volume (@ 5 mg/mL Fe): 5 mL
General Information:
Amine-functionalized Dextran coated crosslinked iron oxide nanoparticles (CLIO-NH2) have 1-3 superparamagnetic Fe3O4 iron oxide cores (8-10 nm in diameter) imbedded within the matrix of dextran sugar strands of 40 kDa or 70 kDa size. Dextran strands are crosslinked to prevent their disassociation, ensuring long-term biological stability. Dextran surface has been functionalized with Amine NH2 groups.
Features:
Highly biocompatible, non-toxic
Long term stable under physiological conditions
40 kDa and 70 kDa dextran coats are available
Highly magnetic
Can be easily conjugated to NHS-activated ligands
Applications:
In vitro and in vivo magnetic labels for cells and tissues
Magnetic cell separation
Probes for MRI
Specifications:
Iron Oxide Core Diameter: 8 – 10 nm
Hydrodyn
Catalog No. NC3593155 Supplier LUNA NANOTECH INC Supplier No. CLIONH2405
Dextran Size: 40 kDa
Volume (@ 5 mg/mL Fe): 5 mL
General Information:
Amine-functionalized Dextran coated crosslinked iron oxide nanoparticles (CLIO-NH2) have 1-3 superparamagnetic Fe3O4 iron oxide cores (8-10 nm in diameter) imbedded within the matrix of dextran sugar strands of 40 kDa or 70 kDa size. Dextran strands are crosslinked to prevent their disassociation, ensuring long-term biological stability. Dextran surface has been functionalized with Amine NH2 groups.
Features:
Highly biocompatible, non-toxic
Long term stable under physiological conditions
40 kDa and 70 kDa dextran coats are available
Highly magnetic
Can be easily conjugated to NHS-activated ligands
Applications:
In vitro and in vivo magnetic labels for cells and tissues
Magnetic cell separation
Probes for MRI
Specifications:
Iron Oxide Core Diameter: 8 – 10 nm
Hydrodyn