Learn More
Medchemexpress LLC Amine-PEG-amine (MW 5000) | 95.0% | 100 MG
SDP
Catalog No. 5000356452
Encompass_Preferred
Amine-PEG-amine (MW 5000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds consisting of two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. The product is intended for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins
  • Intended for research use only
  • Solid appearance
  • White to off-white color
  • Molecular weight of 5000 (average)
  • 95.0% purity
Catalog No. 50-003-56452 Supplier Medchemexpress LLC Supplier No. HY140648100MG
Amine-PEG-amine (MW 5000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds consisting of two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. The product is intended for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins
  • Intended for research use only
  • Solid appearance
  • White to off-white color
  • Molecular weight of 5000 (average)
  • 95.0% purity