Learn More
Medchemexpress LLC Amine-PEG-CH2COOH | 95.0% | 100 MG
SDP
Catalog No. 5000352929
Encompass_Preferred
Amine-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins by facilitating the connection between an E3 ubiquitin ligase and the target protein.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Appears as a solid with a light yellow to yellow color
  • For research use only
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-52929 Supplier Medchemexpress LLC Supplier No. HY140643100MG
Amine-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins by facilitating the connection between an E3 ubiquitin ligase and the target protein.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Appears as a solid with a light yellow to yellow color
  • For research use only
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light