Learn More
Medchemexpress LLC Amine-PEG-CH2COOH (MW 2000) | 139729-28-5 | 95.0% | 50 MG
SDP
Catalog No. 5000353354
Encompass_Preferred
Amine-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker primarily used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). It is designed for research purposes and is not intended for patient use.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-53354 Supplier Medchemexpress LLC Supplier No. HY14064350MG
Amine-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker primarily used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). It is designed for research purposes and is not intended for patient use.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only