Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amine-PEG-thiol (MW 2000) | 93.0% | 100 MG
SDP

Catalog No. 5000356911
Encompass_Preferred

Amine-PEG-thiol (MW 2000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Molecular weight: 2000 (average)
  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: powder -20°C 3 years; in solvent -80°C 6 months, -20°C 1 month
  • Solubility: in DMSO: 50 mg/mL (need ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)
  • For research use only

Catalog No. 50-003-56911 Supplier Medchemexpress LLC Supplier No. HY140651100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amine-PEG-thiol (MW 2000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Molecular weight: 2000 (average)
  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: powder -20°C 3 years; in solvent -80°C 6 months, -20°C 1 month
  • Solubility: in DMSO: 50 mg/mL (need ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.