Learn More
Medchemexpress LLC Amine-PEG-thiol (MW 2000) | 93.0% | 50 MG
SDP
Catalog No. 5000352928
Encompass_Preferred
Amine-PEG-thiol (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound functions by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins, and it appears as a white to off-white solid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • White to off-white solid appearance
Catalog No. 50-003-52928 Supplier Medchemexpress LLC Supplier No. HY14065150MG
Amine-PEG-thiol (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound functions by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins, and it appears as a white to off-white solid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • White to off-white solid appearance