Learn More
Medchemexpress LLC Amine-PEG3-Biotin | 359860-27-8 | 99.8% | 418.55 | 10 MG
SDP
Catalog No. 5000346432
Encompass_Preferred
Amine-PEG3-Biotin is a signal amplification label that contains a biotin group and a terminal primary amine group. It is for research use only.
  • Signal amplification label
  • Contains a biotin group
  • Contains a terminal primary amine group
  • For research use only
Catalog No. 50-003-46432 Supplier Medchemexpress LLC Supplier No. HY11137750MG
Amine-PEG3-Biotin is a signal amplification label that contains a biotin group and a terminal primary amine group. It is for research use only.
  • Signal amplification label
  • Contains a biotin group
  • Contains a terminal primary amine group
  • For research use only