Learn More
Medchemexpress LLC Amine-PEG3-Biotin | 359860-27-8 | 99.8% | 418.55 | 100 MG
SDP
Catalog No. 5000343124
Encompass_Preferred
Amine-PEG3-Biotin is a signal amplification label that contains a biotin group and a terminal primary amine group.
  • Signal amplification label.
  • Contains a biotin group.
  • Contains a terminal primary amine group.
  • Used in electrochemical immunosensors for N6-methyladenosine detection in human cell lines based on a biotin-streptavidin system and silver-SiO2 signal amplification.
Catalog No. 50-003-43124 Supplier Medchemexpress LLC Supplier No. HY11137710MG
Amine-PEG3-Biotin is a signal amplification label that contains a biotin group and a terminal primary amine group.
  • Signal amplification label.
  • Contains a biotin group.
  • Contains a terminal primary amine group.
  • Used in electrochemical immunosensors for N6-methyladenosine detection in human cell lines based on a biotin-streptavidin system and silver-SiO2 signal amplification.