Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amine-PEG3-Biotin | 359860-27-8 | 99.8% | 418.55 | 50 MG
SDP

Catalog No. 5000346433
Encompass_Preferred

Amine-PEG3-Biotin is a signal amplification label that contains a biotin group and a terminal primary amine group, intended for research use only.

  • Contains a biotin group and a terminal primary amine group.
  • Appears as a white to off-white solid.
  • Store at -20°C, protected from light. If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in H2O at 200 mg/mL (477.84 mM) and DMSO at 100 mg/mL (238.92 mM), with sonication required.

Catalog No. 50-003-46433 Supplier Medchemexpress LLC Supplier No. HY111377100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amine-PEG3-Biotin is a signal amplification label that contains a biotin group and a terminal primary amine group, intended for research use only.

  • Contains a biotin group and a terminal primary amine group.
  • Appears as a white to off-white solid.
  • Store at -20°C, protected from light. If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in H2O at 200 mg/mL (477.84 mM) and DMSO at 100 mg/mL (238.92 mM), with sonication required.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.