Learn More
Medchemexpress LLC Amine-PEG3-desthiobiotin | 2237234-71-6 | 99.9% | C18H36N4O5 | 10MG
SDP
Catalog No. 5000260484
Encompass_Preferred
Amine-PEG3-Desthiobiotin is a PEG-based bifunctional linker combining a short PEG3 spacer, a terminal primary amine, and a desthiobiotin affinity tag. It is designed for PROTAC assembly, bioconjugation, and affinity-capture workflows where reversible binding to streptavidin/avidin aids purification and enrichment.
  • Terminal primary amine for efficient coupling to carboxyl or activated ester functionalities.
  • Desthiobiotin tag enables reversible affinity capture with streptavidin/avidin systems.
  • PEG3 spacer improves solubility and reduces steric hindrance between binding partners.
  • High chemical purity suitable for synthetic and analytical applications.
  • Available in multiple small-scale quantities to support method development and optimization.
Catalog No. 50-002-60484 Supplier Medchemexpress LLC Supplier No. HY13472110MG
Amine-PEG3-Desthiobiotin is a PEG-based bifunctional linker combining a short PEG3 spacer, a terminal primary amine, and a desthiobiotin affinity tag. It is designed for PROTAC assembly, bioconjugation, and affinity-capture workflows where reversible binding to streptavidin/avidin aids purification and enrichment.
  • Terminal primary amine for efficient coupling to carboxyl or activated ester functionalities.
  • Desthiobiotin tag enables reversible affinity capture with streptavidin/avidin systems.
  • PEG3 spacer improves solubility and reduces steric hindrance between binding partners.
  • High chemical purity suitable for synthetic and analytical applications.
  • Available in multiple small-scale quantities to support method development and optimization.