Learn More
Medchemexpress LLC Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 | 2244602-35-3 | 98.0% | 750.84 g/mol | C30H58N10O12 | 10 MG
SDP
Catalog No. 5000204383
Encompass_Preferred
Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 is a branched polyethylene glycol (PEG)-based linker intended for use in the synthesis of antibody-drug conjugates (ADCs). It provides an amine reactive handle and multiple azide (N3) termini for bioconjugation, is supplied as a powder for research use, and has a reported molecular weight of 750.84 g/mol and purity of 98.0%.
  • Branched PEG linker with multiple azide termini for orthogonal conjugation
  • Amine reactive handle for straightforward coupling to biomolecules
  • Compatible with click chemistry for ADC assembly
  • High reported purity supports reproducible conjugation
  • Stable when stored under recommended conditions
Catalog No. 50-002-04383 Supplier Medchemexpress LLC Supplier No. HY15367210MG
Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 is a branched polyethylene glycol (PEG)-based linker intended for use in the synthesis of antibody-drug conjugates (ADCs). It provides an amine reactive handle and multiple azide (N3) termini for bioconjugation, is supplied as a powder for research use, and has a reported molecular weight of 750.84 g/mol and purity of 98.0%.
  • Branched PEG linker with multiple azide termini for orthogonal conjugation
  • Amine reactive handle for straightforward coupling to biomolecules
  • Compatible with click chemistry for ADC assembly
  • High reported purity supports reproducible conjugation
  • Stable when stored under recommended conditions