Learn More
Medchemexpress LLC Amine-PEG3-lys(PEG3-N3)-PEG3-N3 | 2244602-35-3 | 98.0% | 750.84 g/mol | C30H58N10O12 | 100 MG
SDP
Catalog No. 5000204450
Encompass_Preferred
Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 is a branched polyethylene glycol (PEG) linker designed for use in the synthesis of antibody-drug conjugates (ADCs). It features an amine terminus and multiple PEG3 spacers terminated with azide (N3) groups to enable orthogonal conjugation strategies.
  • Branched PEG linker with amine and azide functional groups.
  • Designed for ADC synthesis and bioconjugation.
  • Multiple PEG3 spacers increase solubility and flexibility.
  • Enables orthogonal conjugation via amine and azide chemistries.
  • Typical reported purity 98.0%.
Catalog No. 50-002-04450 Supplier Medchemexpress LLC Supplier No. HY153672100MG
Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 is a branched polyethylene glycol (PEG) linker designed for use in the synthesis of antibody-drug conjugates (ADCs). It features an amine terminus and multiple PEG3 spacers terminated with azide (N3) groups to enable orthogonal conjugation strategies.
  • Branched PEG linker with amine and azide functional groups.
  • Designed for ADC synthesis and bioconjugation.
  • Multiple PEG3 spacers increase solubility and flexibility.
  • Enables orthogonal conjugation via amine and azide chemistries.
  • Typical reported purity 98.0%.