Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 | 2244602-35-3 | 98.0% | 750.84 g·mol⁻¹ | C30H58N10O12 | 25 MG
SDP

Catalog No. 5000204311
Encompass_Preferred

Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 is a branched polyethylene glycol (PEG) linker intended for the construction of antibody-drug conjugates (ADCs). It features a primary amine handle and two azide (N3) termini to enable click chemistry and payload attachment, with high purity and defined molecular weight for controlled bioconjugation.

  • Branched PEG linker with amine and azide functional groups.
  • High purity: 98.0%.
  • Molecular weight: 750.84 g·mol⁻¹.
  • Soluble in DMSO at approximately 100 mg/mL; ultrasonic assistance may be required.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).
  • Suitable for click chemistry and payload attachment in ADC construction.

Catalog No. 50-002-04311 Supplier Medchemexpress LLC Supplier No. HY15367225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 is a branched polyethylene glycol (PEG) linker intended for the construction of antibody-drug conjugates (ADCs). It features a primary amine handle and two azide (N3) termini to enable click chemistry and payload attachment, with high purity and defined molecular weight for controlled bioconjugation.

  • Branched PEG linker with amine and azide functional groups.
  • High purity: 98.0%.
  • Molecular weight: 750.84 g·mol⁻¹.
  • Soluble in DMSO at approximately 100 mg/mL; ultrasonic assistance may be required.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).
  • Suitable for click chemistry and payload attachment in ADC construction.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.