Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 | 2244602-35-3 | 98.0% | 750.84 g/mol | C30H58N10O12 | 5 MG
SDP

Catalog No. 5000204446
Encompass_Preferred

This branched polyethylene glycol (PEG) linker contains a terminal amine and two azide (N3) functional groups and is supplied for research use. It is designed for use in bioconjugation workflows, including the synthesis of antibody-drug conjugates (ADCs).

  • Branched PEG linker with terminal amine and azide functionalities.
  • Suitable for antibody-drug conjugate synthesis and other bioconjugations.
  • High purity of 98.0%.
  • Molecular weight 750.84 g/mol.
  • Appearance: yellow to brown oil.
  • Storage: pure form -20°C for 3 years; 4°C for 2 years.

Catalog No. 50-002-04446 Supplier Medchemexpress LLC Supplier No. HY1536725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This branched polyethylene glycol (PEG) linker contains a terminal amine and two azide (N3) functional groups and is supplied for research use. It is designed for use in bioconjugation workflows, including the synthesis of antibody-drug conjugates (ADCs).

  • Branched PEG linker with terminal amine and azide functionalities.
  • Suitable for antibody-drug conjugate synthesis and other bioconjugations.
  • High purity of 98.0%.
  • Molecular weight 750.84 g/mol.
  • Appearance: yellow to brown oil.
  • Storage: pure form -20°C for 3 years; 4°C for 2 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.