Learn More
Medchemexpress LLC Amine-PEG3-lys(PEG3-N3)-PEG3-N3 | 2244602-35-3 | 98.0% | 750.84 g/mol | C30H58N10O12 | 50 MG
SDP
Catalog No. 5000204426
Encompass_Preferred
Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 is a branched polyethylene glycol (PEG)-based linker with an amine terminus and azide functional groups. It is designed for use in antibody-drug conjugate (ADC) synthesis and general bioconjugation workflows where orthogonal azide chemistry and an amine handle are required.
  • Branched PEG linker with amine and azide handles.
  • Suitable for azide-alkyne click chemistry and amine coupling.
  • High purity (98.0%).
  • Available in multiple pack sizes, including 50 MG.
  • Appearance: yellow to brown oil; stored at -20°C (pure) or -80°C in solvent for extended stability.
Catalog No. 50-002-04426 Supplier Medchemexpress LLC Supplier No. HY15367250MG
Amine-PEG3-Lys(PEG3-N3)-PEG3-N3 is a branched polyethylene glycol (PEG)-based linker with an amine terminus and azide functional groups. It is designed for use in antibody-drug conjugate (ADC) synthesis and general bioconjugation workflows where orthogonal azide chemistry and an amine handle are required.
  • Branched PEG linker with amine and azide handles.
  • Suitable for azide-alkyne click chemistry and amine coupling.
  • High purity (98.0%).
  • Available in multiple pack sizes, including 50 MG.
  • Appearance: yellow to brown oil; stored at -20°C (pure) or -80°C in solvent for extended stability.