Learn More
Medchemexpress LLC Amine-PEG6-thiol hydrochloride | 00-00-0 | >98.0% | C14H32ClNO6S | 10MG
SDP
Catalog No. 5000261798
Encompass_Preferred
Amine-PEG6-thiol hydrochloride is a bifunctional PEG6 linker that provides a terminal amine and a terminal thiol for use in PROTAC synthesis and other bioconjugation chemistries. Supplied as the hydrochloride salt, it is a viscous liquid with a molecular weight of 377.92 g/mol and formula C14H32ClNO6S. The material is available in small pack sizes and has storage recommendations to maintain stability.
  • Provides terminal amine and thiol functionalities for dual conjugation.
  • Six-unit polyethylene glycol spacer improves solubility and flexibility.
  • Reported high purity (>98.0%).
  • Viscous liquid form suitable for solution-phase synthesis workflows.
  • Defined molecular weight (377.92 g/mol) and molecular formula for planning reactions.
  • Storage conditions support long-term stability of the pure material.
Catalog No. 50-002-61798 Supplier Medchemexpress LLC Supplier No. HY134711A10MG
Amine-PEG6-thiol hydrochloride is a bifunctional PEG6 linker that provides a terminal amine and a terminal thiol for use in PROTAC synthesis and other bioconjugation chemistries. Supplied as the hydrochloride salt, it is a viscous liquid with a molecular weight of 377.92 g/mol and formula C14H32ClNO6S. The material is available in small pack sizes and has storage recommendations to maintain stability.
  • Provides terminal amine and thiol functionalities for dual conjugation.
  • Six-unit polyethylene glycol spacer improves solubility and flexibility.
  • Reported high purity (>98.0%).
  • Viscous liquid form suitable for solution-phase synthesis workflows.
  • Defined molecular weight (377.92 g/mol) and molecular formula for planning reactions.
  • Storage conditions support long-term stability of the pure material.