Learn More
Biotium Amine Reactive Dyes Reactive CF Dyes Other Dyes Biotinylation Reagents Reactive CF Dyes CF505 1/EA
Catalog No. 5000501407
Encompass_Preferred
CF Dye Succinimidyl Esters (SE NHS esters) are Biotiums next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins CF505 is a green-fluorescent STORM-compatible dye that is structrually equivalent to ATTO 488 Ex/Em 505/519 nm 1 umol per vial
Catalog No. 50-005-01407 Supplier Biotium Supplier No. 97500
CF Dye Succinimidyl Esters (SE NHS esters) are Biotiums next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins CF505 is a green-fluorescent STORM-compatible dye that is structrually equivalent to ATTO 488 Ex/Em 505/519 nm 1 umol per vial