Learn More
Biotium Amine Reactive Dyes Reactive CF Dyes Other Dyes Biotinylation Reagents Reactive CF Dyes CF710 1/EA
Catalog No. 5000501470
Encompass_Preferred
CF Dye Succinimidyl Esters (SE also known as NHS esters) are Biotiums next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups such as lysine residues Near-infrared fluorescent CF710 has excitation/emission at 712/736 nm
Catalog No. 50-005-01470 Supplier Biotium Supplier No. 96125
CF Dye Succinimidyl Esters (SE also known as NHS esters) are Biotiums next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups such as lysine residues Near-infrared fluorescent CF710 has excitation/emission at 712/736 nm