Learn More
Amine-Sulfuric Acid Stock Solution, for Sulfide Determination (Methylene Blue Method), Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Odor | Odorless |
| Physical Form | Liquid |
| Color | Brown |
| pH | < 3 |
| Solubility | Infinitely soluble in water |
| Quantity | 120 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.