Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amine-Sulfuric Acid Stock Solution, for Sulfide Determination (Methylene Blue Method), Ricca Chemical
SDP

Catalog No. R0609900500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R0609900500 500 mL Safety-coated Glass Bottle
R0609900120 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0609900500 Supplier Ricca Chemical Company Supplier No. R0609900500C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Safety-coated Glass Bottle
Odor Odorless
Physical Form Liquid
Color Brown
pH < 3
Solubility Infinitely soluble in water
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.