Learn More
Bio Rad Laboratories AMINEX HPX-87C COLUMN 250X4MM
Catalog No. NC9313279
Encompass
Aminex HPX-87C Columns, 250 andtimes; 4mm, Particle Size: 9andmu;m, Cross linkage: 8%, 5-9pH Range, Ionic Form: Calcium, Applications: sugar alcohols, Packed with a polystyrene divinylbenzene resin
Catalog No. NC9313279 Supplier Bio Rad Laboratories Supplier No. 1250094
Aminex HPX-87C Columns, 250 andtimes; 4mm, Particle Size: 9andmu;m, Cross linkage: 8%, 5-9pH Range, Ionic Form: Calcium, Applications: sugar alcohols, Packed with a polystyrene divinylbenzene resin