Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amino-2-Naphthol-4-Sulfonic Acid, 0.25% w/v APHA 4500, ASTM D859-05, Reagents

Catalog No. 01335891
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-891 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-891 Supplier Reagents Holdings Llc Supplier No. CA215000125C
Add to Cart
Add to Cart

Amino-2-Naphthol-4-Sulfonic Acid, 0.25% w/v APHA 4500, ASTM D859-05, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7631-90-5 , 7631-90-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym ANSA
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 100°C
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.05
Quantity 125 mL
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Amino-2-Naphthol-4-Sulfonic Acid, 0.25% w/v APHA 4500, ASTM D859-05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.