Learn More
Amino-2-Naphthol-4-Sulfonic Acid, 0.25% w/v APHA 4500, ASTM D859-05, Reagents
Amino-2-Naphthol-4-Sulfonic Acid, 0.25% w/v APHA 4500, ASTM D859-05, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$55.30
Chemical Identifiers
| CAS | 7732-18-5 , 7631-90-5 , 7631-90-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | ANSA |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-891
|
Reagents Holdings Llc
CA215000125C |
125 mL |
Each of 1 for $55.30
|
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Chemical Identifiers
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Colorless | |
| 100°C | |
| ANSA | |
| O | |
| 18.02 | |
| Glass Bottle | |
| Non-sterile | |
| Amino-2-Naphthol-4-Sulfonic Acid, 0.25% w/v APHA 4500, ASTM D859-05 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.