Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amino-3,5-dibromopyrazine | 24241-18-7 | 100.0% | C4H3Br2N3 | 500 G
SDP

Catalog No. 5000433096
Encompass_Preferred

Amino-3,5-dibromopyrazine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Solid appearance, color ranges from off-white to yellow.
  • Store at 4°C, protected from light.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Solubility in DMSO is ≥ 100 mg/mL.

Catalog No. 50-004-33096 Supplier Medchemexpress LLC Supplier No. HY79805500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-3,5-dibromopyrazine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Solid appearance, color ranges from off-white to yellow.
  • Store at 4°C, protected from light.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Solubility in DMSO is ≥ 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.