Learn More
Waters Corp Amino Acid Analysis Column, Strong Cation Exchange
Catalog No. 502392280 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This amino acid analysis column is designed for amino acid applications. It is a strong cation exchange column, used for separating and identifying amino acids under conventional chromatographic conditions.
  • Designed for amino acid applications.
  • Strong cation exchange column.
  • Separates and identifies amino acids.
  • Customizable quotes available.
  • Support through documents.
  • Downloadable Certificates of Analysis.
  • Efficiency test reports are an option.
Catalog No. 50-239-2280 Supplier Waters Corp Supplier No. WAT080002
This amino acid analysis column is designed for amino acid applications. It is a strong cation exchange column, used for separating and identifying amino acids under conventional chromatographic conditions.
  • Designed for amino acid applications.
  • Strong cation exchange column.
  • Separates and identifies amino acids.
  • Customizable quotes available.
  • Support through documents.
  • Downloadable Certificates of Analysis.
  • Efficiency test reports are an option.