Learn More
Waters Corp Waters Amino Acid Cell Culture Standard Kit, 26 Amino Acids, Lyophilized Powder
Catalog No. 502038975 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Amino Acid Cell Culture Standard Kit is for identification and quantitative analysis of 26 amino acids in cell culture media and other matrices. It provides lyophilized pre-weighed powder standards for reliable and accurate amino acid determination, supporting various research applications.
  • Contains 26 amino acids, with two vials of 17-hydrolysate standard and eight vials of 9 supplemental cell culture amino acids.
  • Formulated as lyophilized pre-weighed powder for convenient reconstitution.
  • Designed for qualitative and quantitative amino acid analysis using LC systems.
  • Reconstitutes to 1.00 μmol/mL for most amino acids (0.50 μmol/mL for cystine) with 0.1M HCl.
  • Initial storage -10°C to -25°C; stable up to 60 days post-reconstitution (glutamine up to 15 days).
  • Supports method development and troubleshooting.
Catalog No. 50-203-8975 Supplier Waters Corp Supplier No. 186009300
The Amino Acid Cell Culture Standard Kit is for identification and quantitative analysis of 26 amino acids in cell culture media and other matrices. It provides lyophilized pre-weighed powder standards for reliable and accurate amino acid determination, supporting various research applications.
  • Contains 26 amino acids, with two vials of 17-hydrolysate standard and eight vials of 9 supplemental cell culture amino acids.
  • Formulated as lyophilized pre-weighed powder for convenient reconstitution.
  • Designed for qualitative and quantitative amino acid analysis using LC systems.
  • Reconstitutes to 1.00 μmol/mL for most amino acids (0.50 μmol/mL for cystine) with 0.1M HCl.
  • Initial storage -10°C to -25°C; stable up to 60 days post-reconstitution (glutamine up to 15 days).
  • Supports method development and troubleshooting.