Learn More
Hach Company Amino Acid F Dilution Solvent, for Rapid Liquid Silica, 475 mL
Catalog No. 501995867 Shop All Hach Company Products
Encompass
For determination of Ultra Low Range silica by the Heteropoly Blue Rapid Liquid method. Hach method 8282 Range: 3 - 1000 μg/L SiO2. Used with Amino Acid F Reagent Powder (2651155). Rapid Liquid™ methods offer an economical way to analyze multiple samples and to measure trace concentrations. Requires Pour-Thru Cell.
Catalog No. 50-199-5867 Supplier Hach Company Supplier No. 2353011
For determination of Ultra Low Range silica by the Heteropoly Blue Rapid Liquid method. Hach method 8282 Range: 3 - 1000 μg/L SiO2. Used with Amino Acid F Reagent Powder (2651155). Rapid Liquid™ methods offer an economical way to analyze multiple samples and to measure trace concentrations. Requires Pour-Thru Cell.