Learn More
Hach Company Amino Acid F Reagent Powder Pillows, 10 ml, 100/pk
Supplier:  Hach Company 2254069
Encompass
Reagent for Silica determination by the Heteropoly Blue method. Hach Method 8186. 10 ml, 100/pk
Catalog No. 50-465-238