Learn More
Hach Company Amino Acid F Reagent Powder, for Rapid Liquid Silica, 55 g
Supplier:  Hach Company 2651155
Encompass
For determination of Ultra Low Silica by the Heteropoly Blue Method. Hach Method 8282. Used with Amino Acid F Dilution Solvent (2353011) in Rapid Liquid™ method.
Catalog No. 50-199-6435