Learn More
Hach Company Amino Acid F Reagent Solution, 100mL
Supplier:  Hach Company 2386442
Encompass
For determination of Ultra Low Range silica by the Heteropoly Blue method. Hach method 8282 Range: 3 - 1000 μg/L SiO2. 100 mL bottle
Catalog No. 50-199-5904