Learn More
Hach Company Amino Acid F Reagent Solution, 20 Ampules (ultra-low range)
Catalog No. 501995902 Shop All Hach Company Products
Encompass
For determination of Ultra Low Range silica by the Heteropoly Blue method. Hach method 8282 Range: 3 - 1000 μg/L SiO2. Ampuled for increased stability
Catalog No. 50-199-5902 Supplier Hach Company Supplier No. 2386420
For determination of Ultra Low Range silica by the Heteropoly Blue method. Hach method 8282 Range: 3 - 1000 μg/L SiO2. Ampuled for increased stability