Learn More
Waters Corp Amino Acid Internal Standard-Norvaline
Catalog No. NC1810976 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The use of an internal standard best compensates for the variability generated in sample hydrolysis and amino acid analysis. Internal standard norvaline is lyophilized pre-weighed powder. When reconstituted in 6 mL of 0.1M HCl, Norvaline concentration is 0.25 μmol/mL.
Catalog No. NC1810976 Supplier Waters Corp Supplier No. 186009301
The use of an internal standard best compensates for the variability generated in sample hydrolysis and amino acid analysis. Internal standard norvaline is lyophilized pre-weighed powder. When reconstituted in 6 mL of 0.1M HCl, Norvaline concentration is 0.25 μmol/mL.